Learn More
Phospho-Tyrosine Alkaline Phosphatase-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems APM1676
Product Code. 18041722
  • £352.00 / 100µL
Description
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-Tyrosine
Monoclonal
Alkaline Phosphatase (AP)
Supplied as a 0.2 μm filtered solution in phosphate-buffered saline (PBS) with stabilizers and preservatives. with No Preservative
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Detects proteins containing phosphorylated Tyrosine residues. ELISA and 2D Western blot analyses using pervanadate-treated cell lysates indicate that it binds Phospho-Tyrosine in a broad manner largely independent of the surrounding amino acid sequence. This antibody does not cross-react with proteins or peptides containing phosphorylated serine or threonine residues.
Multi-species
Purified
Western Blot, ELISA
179003
Western Blot 1:1000 dilution, ELISA
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine
KLH-coupled Phospho-Tyrosine synthetic peptide.
100 μL
Primary
Sterile PBS to a final concentration of 0.5 mg/mL.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
IgG1