missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tyrosine Agarose-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems AGM1676
This item is not returnable.
View return policy
Description
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Immunoprecipitation.
Specifications
| Phospho-Tyrosine | |
| Monoclonal | |
| Agarose | |
| Phosphate-buffered saline (PBS) with preservative. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects endogenous proteins containing phosphorylated Tyrosine residues. ELISA and 2D Western blot analyses using ligand- and pervanadate-treated cell lysates indicate that clone 179003 binds Phospho-Tyrosine in a broad manner largely independent of the surrounding amino acid sequence. Does not cross-react with proteins or peptides containing phosphorylated serine or threonine residues. | |
| Multi-species | |
| Purified |
| Immunoprecipitation | |
| 179003 | |
| Immunoprecipitation 10 uL/500 ug cell lysate | |
| 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine | |
| KLH-coupled phospho-tyrosine synthetic peptide. | |
| 250 μL | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction