missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-TTK (Thr33, Ser37) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 441325G
This item is not returnable.
View return policy
Description
Phospho-TTK (Thr33, Ser37) Polyclonal Antibody for IP, Western Blot
TTK is a serine/threonine kinase closely related to SPK1, PIM 1 and CDC2 serine/threonine kinases. TTK is a dual-specificity kinase, which can also phosphorylate tyrosine. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. TTK encodes an alpha isoform of the constant regulatory subunit A. Alternatively spliced transcript variants have been described.
Specifications
| Phospho-TTK (Thr33, Ser37) | |
| Polyclonal | |
| Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
| P33981 | |
| TTK | |
| The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human TTK that contains threonine 33 and serine 37. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| TTK | |
| AL022661; cancer/testis antigen 96; CT96; Dual specificity protein kinase TTK; esk; Esk1; monopolar spindle 1 kinase; monopolar spindle 1-like 1; MPH1; MPS1; MPS1L1; OTTHUMP00000016778; OTTHUMP00000220743; phosphotyrosine picked threonine kinase; phosphotyrosine picked threonine-protein kinase; PYT; TTK; Ttk protein kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7272 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction