Learn More
Invitrogen™ Phospho-TRPM7 (Ser1513) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143779
Description
The antibody was cross-adsorbed to unphosphorylated TRPM7 (Ser-1513) peptide then affinity purified using phospho-TRPM7 (Ser-1513) peptide. This antibody detects a 220 kDa* protein on SDS-PAGE immunoblots of human MDA-MB-231 and rat PC12 cells stimulated with calyculin A, as well as detects a 70 kDa autophosphorylated recombinant kinase domain of human TRPM7. These reactivities are not observed after lambda phosphatase treatment to dephosphorylate TRPM7.
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
Specifications
| Phospho-TRPM7 (Ser1513) | |
| Polyclonal | |
| Unconjugated | |
| Trpm7 | |
| 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Channel kinase 1; Channel-kinase 1; FLJ20117; long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250; Ltpr7; LTRPC ion channel family member 7; LTRPC7; LTrpC-7; LTrpC7 {ECO:0000250; PubMed:11161216}; RGD:620053}; tr; transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; Trpm7; trpm7 {ECO:0000312; TRPPLIK; Trp-plik; TRP-PLIK {ECO:0000303; UniProtKB:Q96QT4} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54822, 58800, 679906 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q923J1, Q925B3, Q96QT4 | |
| Trpm7 | |
| Phospho-TRPM7 (Ser-1513) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding Ser-1513 in human TRPM7. This site is well conserved in rat and mouse TRPM7, but has low homology to other TRPM family members. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.