missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-TRF2 (Thr188) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564625
This item is not returnable.
View return policy
Description
Phospho-TRF2 (Thr188) Polyclonal Antibody detects endogenous levels of TRF2 only when phosphorylated at Thr188.
TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.
Specifications
| Phospho-TRF2 (Thr188) | |
| Polyclonal | |
| Unconjugated | |
| TERF2 | |
| Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21750, 361403, 7014 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35144, Q15554 | |
| TERF2 | |
| A synthesized peptide derived from human TRF2 (Phospho-Thr188). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction