missing translation for 'onlineSavingsMsg'
Learn More

Phospho-TPH1 (Ser58) Polyclonal Antibody, Invitrogen™

Product Code. 15555451
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15555451 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15555451 Supplier Invitrogen Supplier No. PA538531

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the aromatic amino acid hydroxylase family. The encoded protein catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. Mutations in this gene have been associated with an elevated risk for a variety of diseases and disorders, including schizophrenia, somatic anxiety, anger-related traits, bipolar disorder, suicidal behavior, addictions, and others.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Tryptophan Hydroxylase (Ser58)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TPH1
Gene Accession No. P17752, P17532, P09810
Gene Alias TPRH, TRPH, L-tryptophan hydroxylase, indoleacetic acid-5-hydroxylase, tryptophan 5-hydroxylase 1, tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase)
Gene Symbols TPH1
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human Tryptophan Hydroxylase around the phosphorylation site of Ser58 (R-N-SP-E-F)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21990, 24848, 7166
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.