missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TLR3 (Tyr759) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564722
This item is not returnable.
View return policy
Description
Phospho-TLR3 (Tyr759) Polyclonal Antibody detects endogenous levels of TLR3 only when phosphorylated at Tyr759.
Phospho-TLR3 (Tyr759) Polyclonal antibody specifically detects Phospho-TLR3 (Tyr759) in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Phospho-TLR3 (Tyr759) | |
| Polyclonal | |
| Unconjugated | |
| TLR3 | |
| AI957183; CD283; IIAE2; TLR 3; TLR3; TLR-3; toll like receptor 3; toll-like receptor 3; toll-like receptor 3 variant 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 142980, 364594, 7098 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15455, Q99MB1 | |
| TLR3 | |
| A synthesized peptide derived from human TLR3 (Phospho-Tyr759) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction