missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-TLK1 (Ser741) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105771
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of TLK1 only when phosphorylated at Ser741.
TLK1 or tousled-like kinase 1 is a nuclear serine/threonine kinase that is potentially involved in the regulation of chromatin assembly. TLK1 is first described in Arabidopsis. TLK1 is a novel target of the DNA damage checkpoint that is rapidly inactivated upon exposure to ionizing radiation (IR) and the inactivation is directly mediated by the S-phase DNA damage checkpoint. The suppression of TLK1 activity after double-strand breaks in the DNA, replication blockade, or low doses of ultraviolet irradiation requires ATM, NBS and CHK1. The Tousled-like kinases, first described in Arabadopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.
Specifications
| Phospho-TLK1 (Ser741) | |
| Polyclonal | |
| Unconjugated | |
| TLK1 | |
| 4930545J15Rik; E(Sp1) homolog; Enhancer of split groucho-like protein 1; ESG; ESG1; GRG1; KIAA0137; PKU beta; PKU-beta; serine threonine protein kinase; serine/threonine-protein kinase tousled-like 1; SNAK; SNARE protein kinase SNAK; TLE1; TLK1; TLK-1; TLK1B; tousled like kinase 1; tousled-like kinase 1; transducin like enhancer of split 1; transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer protein 1 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 228012, 311118, 9874 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C0V0, Q9UKI8 | |
| TLK1 | |
| A synthesized peptide derived from human TLK1(Accession Q9UKI8), corresponding to amino acid residues around phosphorylated Ser741. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction