missing translation for 'onlineSavingsMsg'
Learn More

Phospho-TK1 (Ser13) Polyclonal Antibody, Invitrogen™

Product Code. 15575261
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15575261 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15575261 Supplier Invitrogen Supplier No. PA538340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Thymidine kinase (TK) belongs to a group of enzymes such as dihydrofolate reductase, thymidylate synthase, and DNA polymerase that are involved in DNA synthesis and precursor production. High levels of these enzymes are present in proliferating cells, and low levels are found in resting cells, due to multiple regulatory mechanisms that ensure exclusive expression of these enzymes in replicating cells. TK is responsible for catalyzing the phosphorylation of thymidine, which functions as a part of the pyrimidine salvage pathway involved in DNA synthesis. The activities of enzymes such as TK are essential for the activation of several chemotherapeutically important nucleoside analogues that are administered as prodrugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TK1 (Ser13)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TK1
Gene Accession No. P04183, P04184
Gene Alias EC 2.7.1.21, KITH, TK-1, TK1, Thymidine kinase, cytosolic
Gene Symbols TK1
Host Species Rabbit
Immunogen A synthetic phosphopeptide derived from human TK around the phosphorylation site of Ser13 (P-G-SP-P-S)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21877, 7083
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.