missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-TGFBR3 (Thr842) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564528
This item is not returnable.
View return policy
Description
Phospho-TGFBR3 (Thr842) Polyclonal Antibody detects endogenous levels of TGFBR3 only when phosphorylated at Thr842.
This locus encodes the transforming growth factor (TGF)-beta type III receptor. The encoded receptor is a membrane proteoglycan that often functions as a co-receptor with other TGF-beta receptor superfamily members. Ectodomain shedding produces soluble TGFBR3, which may inhibit TGFB signaling. Decreased expression of this receptor has been observed in various cancers. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications
| Phospho-TGFBR3 (Thr842) | |
| Polyclonal | |
| Unconjugated | |
| TGFBR3 | |
| Betaglycan; betaglycan proteoglycan; TGF-beta receptor type 3; TGF-beta receptor type III; TGFR-3; Transforming growth factor beta receptor III; Transforming growth factor beta receptor type 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21814, 29610, 7049 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q03167, O88393, P26342 | |
| TGFBR3 | |
| A synthesized peptide derived from human TGF beta Receptor III (Phospho-Thr842) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction