missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-TCF3 (Thr355) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537753
This item is not returnable.
View return policy
Description
A suggested positive control for IHC is human brain tissue.
TCF3 (E2A) is a transcription factor that plays major roles in determining tissue-specific cell fate during embryogenesis, like muscle or early B-cell differentiation. Heterodimers between E2A and tissue-specific basic helix-loop-helix (bHLH) Dimers bind DNA on E-box motifs: 5'- CANNTG-3'. Binds to the kappa-E2 site in the kappa immunoglobulin gene enhancer. Deletions in E2A have been observed in a subset of pre-B-cell acute lymphoblastic leukemia (B-ALL) cases. Two alternatively spliced human isoforms have been described.
Specifications
| Phospho-TCF3 (Thr355) | |
| Polyclonal | |
| Unconjugated | |
| TCF3 | |
| A1; AA408400; AGM8; Alf2; AW209082; bHLHb21; class B basic helix-loop-helix protein 21; E12; E12/E47; E2A; E47; helix-loop-helix protein HE47; immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; ITF1; Kappa-E2-binding factor; Me2; MGC129647; MGC129648; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; Pan; Pan1; Pan2; pancreas specific transcription factor 1c; Ptf1c; TCF3; TCF-3; Tcfe2a; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; transcription factor A1; transcription factor E2a; Transcription factor E2-alpha; transcription factor ITF-1; Transcription regulator Pan; VDIR; VDR interacting repressor; vitamin D receptor-interacting repressor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 171046, 21423, 6929 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15806, P15923, P21677 | |
| TCF3 | |
| Peptide sequence around phosphorylation site of threonine 355 (P-S-T(p)-P-V) derived from Human E2A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction