missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-TACC3 (Ser558) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105244
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of TACC3 only when phosphorylated at Ser558.
TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.
Specifications
| Phospho-TACC3 (Ser558) | |
| Polyclonal | |
| Unconjugated | |
| TACC3 | |
| Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 10460, 21335, 360962 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9JJ11, Q9Y6A5 | |
| TACC3 | |
| A synthesized peptide derived from human TACC3(Accession Q9Y6A5), corresponding to amino acid residues around phosphorylated Ser558. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction