missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Syntaxin 1 (Ser14) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105014
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Syntaxin 1A only when phosphorylated at Serine 14.
Syntaxin 1 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE [Soluble NSF (N-ethylmaleimide-sensitive fusion protein)-Attachment protein REceptor] domain (known as H3), and an N-terminal regulatory domain (Habc). Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Phospho-Syntaxin 1 (Ser14) | |
| Polyclonal | |
| Unconjugated | |
| Stx1a | |
| HPC-1; LOW QUALITY PROTEIN: syntaxin-1A; neuron-specific antigen HPC-1; P35-1; P35A; Sap; STX1; Stx1a; STX1B; STX1B1; STX1B2; STXA1; SYN1A; Synaptotagmin-associated 35 kDa protein; syntaxin 1 a; syntaxin 1A; syntaxin 1A (brain); syntaxin 1A (brain)-like; syntaxin-1A; Syntaxin-1B1; Syntaxin-1B2 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 116470, 20907, 6804 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35526, P32851, Q16623 | |
| Stx1a | |
| A synthesized peptide derived from human STX1A(Accession Q16623), corresponding to amino acid residues around phosphorylated Ser14. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction