missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Synapsin 1 (Ser9) Polyclonal Antibody

Phospho-Synapsin 1 (Ser9) Polyclonal Antibody
Brand: Invitrogen PA5104822
This item is not returnable.
View return policy
Description
Phospho-Synapsin 1 (Ser9) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-Synapsin 1 (Ser9) | |
| Polyclonal | |
| Unconjugated | |
| SYN1 | |
| Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 20964, 24949, 6853 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88935, P09951, P17600 | |
| SYN1 | |
| A synthesized peptide derived from human SYN1(Accession P17600), corresponding to amino acid residues around phosphorylated Ser9. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction