Learn More
Invitrogen™ Phospho-Synapsin 1 (Ser549) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14697
Description
This antibody is predicted to react with bovine, canine, human, mouse and non-human primate based on 100% sequence homology. In Western blot, this antibody is specific for ~78 kDa synapsin I doublet phosphorylated at Ser549. Immunolabeling can be blocked by lambda-phosphatase treatment.
SYN1 (synapsin 1) is a member of the synapsin gene family. Synapsins encode neuronal phosphoproteins that associate with the cytoplasmic surface of synaptic vesicles. Family members are characterized by common protein domains and are implicated in synaptogenesis and the modulation of neurotransmitter release, suggesting a potential role in several neuropsychiatric diseases. Synapsin 1 plays a role in regulation of axonogenesis and synaptogenesis. It serves as a substrate for several different protein kinases and phosphorylation may function in the regulation of this protein in the nerve terminal. Mutations in this gene may be associated with X-linked disorders with primary neuronal degeneration such as Rett syndrome. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| Phospho-Synapsin 1 (Ser549) | |
| Polyclonal | |
| Unconjugated | |
| SYN1 | |
| Brain protein 4.1; SYN1; Syn-1; SYN1a; SYN1b; Syn1-S; synapsin 1; synapsin I; synapsin Ia; synapsin Ib; synapsin-1; SYNI; truncated synapsin 1-S | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20964, 24949 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O88935, P09951 | |
| SYN1 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser549 of rat SYN1 conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.