Learn More
Invitrogen™ Phospho-Syk (Tyr348) Monoclonal Antibody (moch1ct), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£113.00 - £267.00
Specifications
| Antigen | Phospho-Syk (Tyr348) |
|---|---|
| Clone | moch1ct |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15537156
|
Invitrogen™
12-9014-41 |
25 Tests |
£113.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15547156
![]() |
Invitrogen™
12-9014-42 |
100 Tests |
£267.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This moch1ct monoclonal antibody recognizes human and mouse spleen tyrosine kinase (also known as SYK) when phosphorylated on tyrosine 348 (Y348). SYK is the founding member of the SYK family of kinases that also includes ZAP-70 (zeta-associated protein of 70 kD) and is expressed in hematopoietic cells, including B lymphocytes, immature (CD4, CD8 double-negative and double-positive) thymocytes, and myeloid cells, epithelial cell lines, and normal breast tissue. SYK is critical for B cell receptor (BCR) signaling and B cell development. Autophosphorylation of at Y348 is necessary for SYK to become fully catalytically active and creates a docking site for the SH2 domains of Vav, Grb2, p85 subunit of PI3 kinase, and PLC gamma. Specificity of this moch1ct clone was confirmed by ELISA, flow cytometry, and western blotting. Applications Reported: This moch1ct antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This moch1ct antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of stimulated mouse splenocytes. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Spleen tyrosine kinase (SYK), along with ZAP70, are cytoplasmic tyrosine kinases that are viewed as potential druge targets for allergic disorders and autoimmune diseases. SYK may also play a role in cancer. Syk translocates to the plasma membrane upon B cell antigen receptor (BCR) or the high-affinity IgE receptor (FcepsilonRI) triggering, and phosphorylates downstream adaptor proteins, thereby providing docking sites for initiation of subsequent signaling pathways, such as calcium mobilization, cytoskeleton remodeling, or transcription of specific genes. Syk binds to the receptor assemblies through interactions of its pair of SH2 domains with ITAM motives of the receptor, which have been phosphorylated by Src-family kinases. These kinases also help to activate Syk by phosphorylation of its activation loop.Specifications
| Phospho-Syk (Tyr348) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK | |
| Syk | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Syk |
| moch1ct | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human, Mouse | |
| P43405, P48025 | |
| 20963, 6850 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.