missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-STYK1 (Tyr327) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564854
This item is not returnable.
View return policy
Description
Phospho-STYK1 (Tyr327) Polyclonal Antibody detects endogenous levels of STYK1 only when phosphorylated at Tyr327.
STYK1, a probable tyrosine protein-kinase, which has strong transforming capabilities on a variety of cell lines. When overexpressed, it can also induce tumor cell invasion as well as metastasis in distant organs. May act by activating both MAP kinase and phosphatidylinositol 3'-kinases (PI3K) pathways. It is widely expressed; highly expressed in brain, placenta and prostate. STYK1 is expressed in tumor cells such as hepatoma cells LO2, cervix carcinoma cells HeLa, ovary cancer cells Ho8910 and chronic myelogenous leukemia cells K562, but not in other tumor cells such as epidermoid carcinoma.
Specifications
| Phospho-STYK1 (Tyr327) | |
| Polyclonal | |
| Unconjugated | |
| STYK1 | |
| 9130025L13; AI326477; DKFZp761P1010; mNOK; Nok; NOK kinase; novel oncogene with kinase domain; Protein PK-unique; RGD1564211; Serine/threonine/tyrosine kinase 1; Styk1; SuRTK106; tyrosine-protein kinase STYK1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 243659, 55359 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6J9G0, Q6J9G1 | |
| STYK1 | |
| A synthesized peptide derived from human STYK1/NOK (Phospho-Tyr327). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction