missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-STK38/STK38L (Thr444, Thr442) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599570
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of NDR1/2 only when phosphorylated at Thr444/442.
This gene encodes a member of the AGC serine/threonine kinase family of proteins. The kinase activity of this protein is regulated by autophosphorylation and phosphorylation by other upstream kinases. This protein has been shown to function in the cell cycle and apoptosis. This protein has also been found to regulate the protein stability and transcriptional activity of the MYC oncogene. Alternative splicing results in multiple transcript variants.
Specifications
| Phospho-STK38/STK38L (Thr444, Thr442) | |
| Polyclonal | |
| Unconjugated | |
| Stk38 | |
| 4930473A22Rik; 5830476G13Rik; 9530097A09Rik; AA617404; B230328I19; KIAA0965; NDR; Ndr Ser/Thr kinase-like protein; NDR1; NDR1 protein kinase; NDR2; NDR2 protein kinase; Ndr54; nuclear Dbf2-related 2; nuclear Dbf2-related kinase 1; Nuclear Dbf2-related kinase 2; RGD1564793; serine/threonine kinase 38; serine/threonine kinase 38 like; serine/threonine kinase 38-like; serine/threonine-protein kinase 38; Serine/threonine-protein kinase 38-like; stk38; Stk38l | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 106504, 11329, 23012, 232533, 361813, 691337 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15208, Q7TSE6, Q91VJ4, Q9Y2H1 | |
| Stk38, STK38L | |
| A synthesized peptide derived from human STK38(Accession Q15208), corresponding to amino acid residues around phosphorylated Thr444. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction