Learn More
Phospho-STK11 (Thr189) Rabbit anti-Human, CoraLite 488, Clone: 5J9, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL488-80127-100UL
Description
The Phospho-STK11 (Thr189) antibody from Proteintech is a rabbit recombinant antibody to a peptide of human Phospho-STK11 (Thr189). This antibody recognizes human, mouse antigen. The Phospho-STK11 (Thr189) antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
| Phospho-STK11 (Thr189) | |
| 5J9 | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 6794 | |
| IgG |