missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Stat4 (Tyr693) Rabbit anti-Human, Mouse, PE, Clone: Stat4Y693-F6, Invitrogen™
Rabbit Monoclonal Antibody
£143.00
Specifications
| Antigen | Phospho-Stat4 (Tyr693) |
|---|---|
| Clone | Stat4Y693-F6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17186918
|
Thermo Scientific
MA537332 |
10 Tests | |||||||
Description
Phospho-Stat4 (Tyr693) Monoclonal antibody specifically detects Phospho-Stat4 (Tyr693) in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| Phospho-Stat4 (Tyr693) | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human, Mouse | |
| P42228, Q14765 | |
| 20849, 6775 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| Stat4Y693-F6 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| CIS4; HSPC060; OTTHUMP00000163553; Signal transducer and activator of transcription 4; SLEB11; SOCS4; SOCS6; SSI4; STAI4; STAT; Stat4; STATI4 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr693 of human phospho Stat4. | |
| Primary | |
| 2-8°C | |
| STAT4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title