missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT4 (Tyr693) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 717900
Code : 50
Additional Details : Weight : 0.50000kg
Description
Phospho-STAT4 (Tyr693) Polyclonal antibody specifically detects Phospho-STAT4 (Tyr693) in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Phospho-STAT4 (Tyr693) | |
Polyclonal | |
Unconjugated | |
STAT4 | |
CIS4; HSPC060; OTTHUMP00000163553; Signal transducer and activator of transcription 4; SLEB11; SOCS4; SOCS6; SSI4; STAI4; STAT; Stat4; STATI4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20849, 6775 | |
-20°C | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P42228, Q14765 | |
STAT4 | |
A synthetic tyrosine phosphorylated peptide encompassing and phosphorylated at the conserved tyrosine phosphorylation site (Y-693) of the murine STAT4 protein. | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |