missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-STAT2 (Tyr690) Polyclonal Antibody
GREENER_CHOICE

Product Code. 11572540
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11572540 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11572540 Supplier Invitrogen™ Supplier No. PA517647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

STAT2 is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. In response to interferon (IFN), this protein forms a complex with STAT1 and IFN regulatory factor family protein p48 (ISGF3G), in which this protein acts as a transactivator, but lacks the ability to bind DNA directly. Transcription adaptor P300/CBP (EP300/CREBBP) has been shown to interact specifically with this protein, which is thought to be involved in the process of blocking IFN-alpha response by adenovirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-STAT2 (Tyr690)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Stat2
Gene Accession No. P52630
Gene Alias 1110034C02Rik; 1600010G07Rik; acute phase response factor; acute-phase response factor; ADMIO; ADMIO1; Aprf; AW109958; AW496480; DNA-binding protein APRF; EGK_03803; HIES; IMD44; interferon alpha induced transcriptional activator; ISGF-3; MGC59816; p113; signal transducer and activator of transcription 2; signal transducer and activator of transcription 2, 113kDa; signal transducer and activator of transcription 3; signal transducer and activator of transcription 3 (acute-phase response factor); STAT; STAT 2; STAT113; STAT2; STAT-2; Stat3
Gene Symbols Stat2
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pTyr690 of human Stat2 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288774, 511023, 6773
Target Species Bovine, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.