missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT1 (Y701) Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC2894T-100UG
This item is not returnable.
View return policy
Description
STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| STAT1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91 | |
| Phosphopeptide containing the human STAT1 Y701 site, Accession # NP009330 | |
| 100 μg | |
| Primary | |
| Detects human STAT1 when phosphorylated at Y701 in Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
| Flow Cytometry | |
| 1086B | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP009330 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6772 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction