missing translation for 'onlineSavingsMsg'
Learn More

Phospho-SMAD2/SMAD3 (Ser465, Ser467, Ser423, Ser425) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA5-110155

Product Code. 16695884

  • £355.04 / 100µL
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: SSMS

Smad proteins, the mammalian homologs of the Drosophila mothers against decapentaplegic (Mad), have been implicated as downstream effectors of TGFβ/BMP signaling. Smad1 (also designated Madr1 or JV4-1) and Smad5 are effectors of BMP-2 and BMP-4 function, while Smad2 (also designated Madr2 or JV18-1) and Smad3 are involved in TGFβ and Activin-mediated growth modulation. Smad4 (also designated DPC4) has been shown to mediate all of the above activities through interaction with various Smad family members. Smad6 and Smad7 regulate the response to Activin/TGFβ signaling by interfering with TGFβ-mediated phosphorylation of other Smad proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-SMAD2/SMAD3 (Ser465, Ser467, Ser423, Ser425)
Polyclonal
Unconjugated
SMAD3
7120426M23Rik; AU022421; hMAD-2; hSMAD2; HSPC193; HsT17436; JV15-2; JV18; JV18-1; LDS1C; LDS3; MAD homolog 2; MAD homolog 3; Mad protein homolog; mad3; MADH2; MADH3; Madr2; Mad-related protein 2; mMad2; Mothers against decapentaplegic homolog 2; Mothers against decapentaplegic homolog 3; pSMAD2; SMAD; SMAD 2; SMAD 3; SMAD family member 2; SMAD family member 3; Smad2; Smad-2; Smad3
Rabbit
Antigen affinity chromatography
RUO
4087, 4088
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
0.58 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P84022, Q15796
SMAD2, SMAD3
A synthetic phosphorylated peptide around S465 & S467 of human Smad2 (NP_005892.1).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.