missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SMAD1 (Ser214) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564514
This item is not returnable.
View return policy
Description
Phospho-SMAD1 (Ser214) Polyclonal Antibody detects endogenous levels of SMAD1 only when phosphorylated at Ser214.
The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed.
Specifications
| Phospho-SMAD1 (Ser214) | |
| Polyclonal | |
| Unconjugated | |
| SMAD1 | |
| AI528653; BSP1; BSP-1; dwarfin-A; Dwf-A; DWFC; hSMAD1; JV41; JV4-1; JV5-1; MAD (mothers against decapentaplegic, Drosophila) homolog 1; MAD homolog 1; MAD homolog1 (mothers against decapentaplegic, Drosophila); MAD, mothers against decapentaplegic homolog 1; Madh1; MADH5; MADR1; Mad-related protein 1; Mlp1; mMad1; mothers against decapentaplegic homolog 1; mothers against DPP homolog 1; mothers-against-DPP-related 1; Mothers-against-DPP-related-1; MusMLP; OTTHUMP00000220456; SMAD; Smad 1; SMAD family member 1; SMAD, mothers against DPP homolog 1; Smad1; SMAD5; TGF-beta signaling protein 1; transforming growth factor-beta signaling protein 1; Transforming growth factor-beta-signaling protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17125, 25671, 4086 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70340, P97588, Q15797 | |
| SMAD1 | |
| A synthesized peptide derived from human Smad1 (Phospho-Ser214). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction