missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SKI (Ser383) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105311
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of SKI only when phosphorylated at Ser383.
TGF-beta is a ubiquitously expressed cytokine that signals through the Smad protein family to regulate numerous cellular processes such as embryonic development and tumorigenesis. This signaling is negatively regulated by Ski, the mammalian homolog of the transforming protein of an avian retrovirus that induces oncogenic transformation of chicken embryo cells, and the related protein SnoN. Ski functions by binding to the Smad proteins and preventing their phosphorylation, thereby inhibiting their ability to bind DNA and activate the transcription of downstream genes. Ski will also bind to the Smad proteins specific to bone morphogenic proteins (BMPs) and block BMP signaling in mammalian cells.
Specifications
| Phospho-SKI (Ser383) | |
| Polyclonal | |
| Unconjugated | |
| SKI | |
| 2310012I02Rik; 2610001A11Rik; AA062172; AA589460; BC004088; C-ski; Proto-oncogene c-Ski; RGD1565591; SGS; SKI; Ski oncogene; ski oncoprotein; SKI proto-oncogene; ski sarcoma viral oncogene homolog; ski sarcoma viral oncogene homolog (avian); SKV; Sloan-Kettering viral oncogene homolog; v-ski avian sarcoma viral oncogene homolog | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 20481, 313757, 6497 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12755, Q60698 | |
| SKI | |
| A synthesized peptide derived from human SKI(Accession P12755), corresponding to amino acid residues around phosphorylated Ser383. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction