missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SHP-1 (Tyr536) Recombinant Rabbit Monoclonal Antibody (Shp1Y536-2A7), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£185.00 - £462.00
Specifications
| Antigen | Phospho-SHP-1 (Tyr536) |
|---|---|
| Clone | Shp1Y536-2A7 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17116968
|
Invitrogen™
MA537375 |
10 Tests |
£185.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17156718
![]() |
Invitrogen™
MA537129 |
100 Tests |
£462.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHP-1 (PTPN6) is a non-receptor protein tyrosine phosphatase that is expressed primarily in hematopoietic cells. The enzyme is composed of two SH2 domains, a tyrosine phosphatase catalytic domain, and a carboxy-terminal regulatory domain. SHP-1 removes phosphates from target proteins to downregulate several tyrosine kinase-regulated pathways. In hematopoietic cells, the amino-terminal SH2 domain of SHP-1 binds to tyrosine phosphorylated erythropoietin receptors (EpoR) to negatively regulate hematopoietic growth. Overexpression of SHP-1 in epithelial cells results in dephosphorylation of the Ros receptor tyrosine kinase and subsequent downregulation of Ros-dependent cell proliferation and transformation. Following ligand binding in myeloid cells, SHP-1 associates with the IL-3R beta chain and downregulates IL-3-induced tyrosine phosphorylation and cell proliferation.Specifications
| Phospho-SHP-1 (Tyr536) | |
| Flow Cytometry | |
| PE | |
| Rabbit | |
| Human | |
| P29350 | |
| 5777 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr536 of human phospho Shp1. | |
| Primary | |
| 4°C | |
| PTPN6 |
| Shp1Y536-2A7 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 70Z-SHP; EC 3.1.3.48; hcp; Hcph; hematopoietic cell phosphatase; hematopoietic cell protein-tyrosine phosphatase; HPTP1C; me; motheaten; protein tyrosine phosphatase, non-receptor type 6; protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTN6; Ptp1C; PTP-1C; Ptph6; PTPN6; PTPTY-42; SH2 phosphatase 1; SHP1; Shp-1; SHP-1L; SH-PTP1; Tyrosine-protein phosphatase non-receptor type 6 | |
| PTPN6 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title