missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-SHIP2 (Tyr986, Tyr987) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5104652
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of INPPL1 only when phosphorylated at Tyr986/987.
The protein encoded by this gene is an SH2-containing 5'-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodeling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.
Specifications
| Phospho-SHIP2 (Tyr986, Tyr987) | |
| Polyclonal | |
| Unconjugated | |
| INPPL1 | |
| 51C; 51C protein; ablSH3-binding protein; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; inositol polyphosphate phosphatase-like protein 1; Inppl1; INPPL-1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C; SH2 domain-containing inositol 5'-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5'-phosphatase 2; SH2-containing inositol phosphatase 2; SHIP2; SHIP-2 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 16332, 3636, 65038 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15357, Q6P549, Q9WVR3 | |
| INPPL1 | |
| A synthesized peptide derived from human INPPL1(Accession O15357), corresponding to amino acid residues around phosphorylated Tyr986. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction