missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-(Ser/Thr) ATM + ATR Substrate Polyclonal Antibody
GREENER_CHOICE

Product Code. 17209733 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17209733 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17209733 Supplier Invitrogen™ Supplier No. PA5117231

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, NIH/3T3, C6.

Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. The identified substrates for ATM are p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The essential requirement for the substrates of ATM/ATR is S*/T*Q. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate recognition by these kinases. Positively charged residues surrounding the S*/T*Q are negative determinants for substrate phosphorylation. The complex phenotype of AT cells suggests that it likely has additional substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-(Ser/Thr) ATM + ATR Substrate
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias L(s/t)Q; SQ/TQ
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence containing phosphorylated S & T.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.