missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SEMA4B (Ser809) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105373
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Semaphorin 4B only when phosphorylated at Ser809.
Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. [UniProt].
Specifications
| Phospho-SEMA4B (Ser809) | |
| Polyclonal | |
| Unconjugated | |
| SEMA4B | |
| KIAA1745; mKIAA1745; sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Sema4b; SEMAC; semaphorin 4B; semaphorin-4B; Semaphorin-C; SemC; UNQ749/PRO1480 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 10509, 20352 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q62179, Q9NPR2 | |
| SEMA4B | |
| A synthesized peptide derived from human SEMA4B(Accession Q9NPR2), corresponding to amino acid residues around phosphorylated Ser809. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction