missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-S6 Ribosomal protein (Ser240/244) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81184-1-RR-20UL
This item is not returnable.
View return policy
Description
The Phospho-S6 Ribosomal protein (Ser240/244) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-S6 Ribosomal protein (Ser240/244). This antibody recognizes human antigen. The Phospho-S6 Ribosomal protein (Ser240/244) antibody has been validated for the following applications: WB, IF/ICC, FC (Intra), ELISA analysis.Specifications
| S6 Ribosomal protein (Ser240/244) | |
| Recombinant Monoclonal | |
| 400 μg/mL | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 6194 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 5J13 | |
| Unconjugated | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| 32 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction