missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RyR2 (Ser2808) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105712
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of RYR2 only when phosphorylated at Ser2808.
Dihydropyridine receptor (DHPR) is a surface membrane protein critical for the excitation-contraction coupling of striated muscle. DHPR and the sarcoplasmic reticulum ryanodine receptor (RyR) are two key components of the intracellular junctions, where depolarization of the surface membrane is converted into the release of Ca2+ from internal stores. The alpha1-subunit of the DHPR contains a cytoplasmic loop which is thought to be involved in the interactions with RyR. Phosphorylation of the DHPR alpha1-subunit is also thought to play a role in the functional interaction of DHPR and RyR. Mutation in DHPR alpha1 results in excitation-contraction uncoupling, leading to muscular dysgenesis, a complete inactivity in developing skeletal muscles. Cells that do not express RyR also lack excitation-contraction coupling and exhibit a severalfold reduction in Ca2+ current density.
Specifications
| Phospho-RyR2 (Ser2808) | |
| Polyclonal | |
| Unconjugated | |
| Ryr2 | |
| 9330127I20Rik; ARVC2; ARVD2; calcium release channel; cardiac muscle ryanodine receptor; Cardiac muscle ryanodine receptor-calcium release channel; cardiac ryanodine receptor 2; cardiac-type ryanodine receptor; hRYR-2; islet-type ryanodine receptor; kidney-type ryanodine receptor; Ryanodine receptor; ryanodine receptor 2; ryanodine receptor 2 (cardiac); ryanodine receptor 2, cardiac; ryanodine receptor type 2; ryanodine receptor type II; RyR; RYR2; RYR-2; Type 2 ryanodine receptor; VTSIP | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 20191, 6262, 689560 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B0LPN4, E9Q401, Q92736 | |
| Ryr2 | |
| A synthesized peptide derived from human RYR2(Accession Q92736), corresponding to amino acid residues around phosphorylated Ser2808. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction