missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RSK3 (Ser360) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5118663
This item is not returnable.
View return policy
Description
Phospho-RSK3 (Ser360) Polyclonal antibody specifically detects Phospho-RSK3 (Ser360) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
| Phospho-RSK3 (Ser360) | |
| Polyclonal | |
| Unconjugated | |
| RPS6KA2 | |
| 90 kDa ribosomal protein S6 kinase 2; 90kDa; D17Wsu134e; HU-2; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1c; MAPKAP kinase 1C; MAPKAPK1C; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; p90rsk; p90-RSK 2; p90RSK2; p90-RSK3; pp90rsk; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; ribosomal S6 kinase 3; Rps6ka2; Rps6ka-rs1; RSK; Rsk3; RSK-3; S6K-alpha; S6K-alpha 2; S6K-alpha2; S6K-alpha-2 | |
| A synthesized peptide derived from human RSK3 around the phosphorylation site of Ser360. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15349, Q9WUT3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117269, 20112, 6196 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction