missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-RSK1 (Thr359, Ser363) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16634255
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16634255 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16634255 Supplier Invitrogen™ Supplier No. PA5101020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of RSK1 p90 only when phosphorylated at PT359+S363.

RPS6KA1 (RSK1) is a growth-factor regulated serine/threonine kinase that is involved in the MAPK cascade. It is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors. RPS6KA1 contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RSK1 (Thr359, Ser363)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPS6KA1
Gene Accession No. P18653, Q15418, Q63531
Gene Alias 90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAPK-1; mapkapk1a; MAPKAPK-1a; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; RPS6KA; Rps6ka1; rps6ka1.L; RPS6KA1L; rsk; RSK1; Rsk-1; S6 kinase II beta; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha-1; S6KII-alpha; S6KII-beta; wu:fc56h07; XELAEV_18012109mg; zgc:152654
Gene Symbols RPS6KA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPS6KA1(Accession Q15418), corresponding to amino acid residues around phosphorylated Thr359.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20111, 6195, 81771
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.