missing translation for 'onlineSavingsMsg'
Learn More

Phospho-RPS6KA1 (Thr359/Ser363) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£125.00 - £280.00

Specifications

Antigen Phospho-RPS6KA1 (Thr359/Ser363)
Concentration 200 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17251436
View Documents
Proteintech
29225-1-AP-20UL
20 μL
£125.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17241436
View Documents
Proteintech
29225-1-AP-100UL
100 μL
£280.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 90 kDa ribosomal S6 kinases (RSK1-4) are a family of widely expressed serine/threonine kinases and play an important role in the MAPK signaling cascade and act downstream of ERK1/2 signaling. It can mediate mitogenic and stress-induced activation of the transcription factors CREB1, ETV1/ER81 and NR4A1/NUR77. RSK1 (RPS6KA1) is also designated as p90RSK associated with cellular proliferation, survival and differentiation. RPS6KA1 has some phosphorylation sites including Ser380, Thr359 and Ser363 which are important for kinase activation (PMID: 16458888, 30813401, 17259979, 10679322)

Specifications

Specifications

Phospho-RPS6KA1 (Thr359/Ser363)
Western Blot, ELISA
Unconjugated
Rabbit
6195
200 μg/mL
Polyclonal
Liquid
Human, Mouse
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.