missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RIP1 (Ser166) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121351
This item is not returnable.
View return policy
Description
Positive test controls include: HT-29, NIH/3T3. Immunogen sequence: MWSKL.
Serine-threonine kinase which transduces inflammatory and cell-death signals (programmed necrosis) following death receptors ligation, activation of pathogen recognition receptors (PRRs), and DNA damage. Upon activation of TNFR1 by the TNF-alpha family cytokines, TRADD and TRAF2 are recruited to the receptor. Phosphorylates DAB2IP at 'Ser-728' in a TNF-alpha-dependent manner, and thereby activates the MAP3K5-JNK apoptotic cascade. Ubiquitination by TRAF2 via 'Lys-63'-link chains acts as a critical enhancer of communication with downstream signal transducers in the mitogen-activated protein kinase pathway and the NF-kappa-B pathway, which in turn mediate downstream events including the activation of genes encoding inflammatory molecules. Polyubiquitinated protein binds to IKBKG/NEMO, the regulatory subunit of the IKK complex, a critical event for NF-kappa-B activation. Interaction with other cellular RHIM-containing adapters initiates gene activation and cell death. RIPK1 and RIPK3 association, in particular, forms a necrosis-inducing complex.
Specifications
| Phospho-RIP1 (Ser166) | |
| Polyclonal | |
| Unconjugated | |
| RIPK1 | |
| cell death protein RIP; D330015H01Rik; FLJ39204; OTTHUMP00000015955; receptor (TNFRSF)-interacting serine-threonine kinase 1; receptor interacting serine/threonine kinase 1; receptor-interacting protein 1; receptor-interacting protein kinase 1; receptor-interacting serine/threonine-protein kinase 1; Rinp; RIP; RIP1; RIP-1; RIP1 (CT); RIPK1; serine/threonine-protein kinase RIP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19766, 8737 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13546, Q60855 | |
| RIPK1 | |
| A synthetic phosphorylated peptide around S166 of human RIPK1/RIP (NP_003795.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction