missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RelB (Ser552) Recombinant Rabbit Monoclonal Antibody (RelBS552-A7), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£185.00 - £463.00
Specifications
| Antigen | Phospho-RelB (Ser552) |
|---|---|
| Clone | RelBS552-A7 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17186888
|
Invitrogen™
MA537302 |
10 Tests |
£185.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17186618
![]() |
Invitrogen™
MA537032 |
100 Tests |
£463.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NFkB transcription factor was originally identified as a protein complex consisting of a DNA-binding subunit and an associated protein. The DNAbinding subunit is functionally related to c-Rel p75 and Rel B p68. The p50 subunit was initially believed to be a functionally unique protein derived from the amino-terminus of a precursor designated p105. A second protein designated p52 (previously referred to as p49) has been identified that can act as an alternative NFkB subunit. Rel B does not bind with high affinity to NFkB sites, but heterodimers between Rel B and p50 bind with an affinity comparable to that of p50 NFkB homodimers. However, Rel B/p50 heterodimers, in contrast to NFkB heterodimers, transactivates transcription of promotors containing kB binding sites.Specifications
| Phospho-RelB (Ser552) | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Human, Mouse | |
| Q01201, Q04863 | |
| 19698, 5971 | |
| A synthetic phospho-peptide corresponding to residues surrounding Ser552 of human phospho RelB. | |
| Primary | |
| 4°C, store in dark | |
| RELB |
| RelBS552-A7 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| avian reticuloendotheliosis viral (v-rel) oncogene related B; IREL; I-REL; OTTHUMP00000221313; rel b; RELB; REL-B; RELB proto-oncogene, NF-kB subunit; shep; transcription factor RelB; v-rel avian reticuloendotheliosis viral oncogene homolog B; v-rel avian reticuloendotheliosis viral oncogene homolog B (nuclear factor of kappa light polypeptide gene enhancer in B-cells 3); v-rel reticuloen; v-rel reticuloendotheliosis viral oncogene homolog B; v-rel reticuloendotheliosis viral oncogene homolog B, nuclear factor of kappa light polypeptide gene enhancer in B-cells 3 | |
| RELB | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title