missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RBL1 (Thr369) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538423
This item is not returnable.
View return policy
Description
RB-like 1 is similar in sequence and possibly function to the product of the retinoblastoma 1 (RB1) gene. The RB1 gene product is a tumor suppressor protein that appears to be involved in cell cycle regulation, as it is phosphorylated in the S to M phase transition and is dephosphorylated in the G1 phase of the cell cycle. Both the RB1 protein and RB-like 1 can form a complex with adenovirus E1A protein and SV40 large T-antigen, with the SV40 large T-antigen binding only to the unphosphorylated form of each protein. In addition, both proteins can inhibit the transcription of cell cycle genes containing E2F binding sites in their promoters. Due to the sequence and biochemical similarities with the RB1 protein, it is thought that RB-like 1 may also be a tumor suppressor.
Specifications
| Phospho-RBL1 (Thr369) | |
| Polyclonal | |
| Unconjugated | |
| RBL1 | |
| Retinoblastoma-like protein 1, 107 kDa retinoblastoma-associated protein, PRB1, P107, RBL1, | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19650, 5933 | |
| -20°C | |
| Liquid |
| Peptide ELISA | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P28749, Q64701 | |
| RBL1 | |
| A synthetic phosphopeptide derived from human RBL1 around the phosphorylation site of Thr369 (P-S-TP-P-L) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction