missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Rb (Ser811) Polyclonal Antibody

Phospho-Rb (Ser811) Polyclonal Antibody
Brand: Invitrogen PA597350
This item is not returnable.
View return policy
Description
Phospho-Rb (Ser811) Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Phospho-Rb (Ser811) | |
| Polyclonal | |
| Unconjugated | |
| RB1 | |
| Cleaved Rb; delta RB-p70; DRb-p70; exon 17 tumor GOS561 substitution mutation causes premature stop; GOS563 exon 17 substitution mutation causes premature stop; OSRC; p104; p104 chicken Rb; P105RB; p105-Rb; Phospho-Retinoblastoma; pp105; pp110; PPP1R130; pRb; prepro-retinoblastoma-associated protein; protein phosphatase 1, regulatory subunit 130; rb; RB transcriptional corepressor 1; RB1; Rb-1; Rb-p70; Retin; Retinoblastoma; retinoblastoma 1; retinoblastoma 1 (including osteosarcoma); retinoblastoma 1 protein; retinoblastoma gene; retinoblastoma isoform A; retinoblastoma protein; retinoblastoma suspectibility protein; retinoblastoma tumor suppressor; retinoblastoma-associated protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5925 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P06400 | |
| RB1 | |
| A synthetic phosphorylated peptide around S811 of human RB (NP_0003122). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction