missing translation for 'onlineSavingsMsg'
Learn More

Phospho-RARA/RARB (Ser96) Polyclonal Antibody, Invitrogen™

Product Code. 16652375
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16652375 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16652375 Supplier Invitrogen Supplier No. PA5105934

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Retinoic Acid Receptor alpha/beta only when phosphorylated at Retinoic AcSer96.

Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-RARA/RARB (Ser96)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RARB
Gene Accession No. P10276, P10826, P11416, P22605
Gene Alias A830025K23, HAP, HBV-activated protein, hepatitis B virus activated protein, MCOPS12, Nr1b1, Nr1b2, Nuclear receptor subfamily 1 group B member 1, nuclear receptor subfamily 1 group B member 2, nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form, RAR, RAR alpha 1, RARA, RAR-alpha, RARalpha1, Rarb, RAR-beta, RARbeta1, RAR-epsilon, retinoic acid, retinoic acid nuclear receptor alpha variant 1, retinoic acid nuclear receptor alpha variant 2, retinoic acid receptor, retinoic acid receptor alpha, Retinoic acid receptor beta, retinoic acid receptor, alpha, retinoic acid receptor, alpha polypeptide, retinoic acid receptor, beta, retinoic acid receptor, beta polypeptide, RRB2
Gene Symbols RARA, RARB
Host Species Rabbit
Immunogen A synthesized peptide derived from human RARA(Accession P10276), corresponding to amino acid residues around phosphorylated Ser96.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19401, 218772, 24705, 24706, 5914, 5915
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.