missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Raptor (Ser792) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517116
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Specifications
| Phospho-Raptor (Ser792) | |
| Polyclonal | |
| Unconjugated | |
| RPTOR | |
| 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Rap; RAPTOR; regulatory associated protein of MTOR complex 1; regulatory associated protein of MTOR, complex 1; regulatory-associated protein of mTOR; RGD1311784; RPTOR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 287871, 57521, 74370 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 52 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q8K4Q0, Q8N122 | |
| RPTOR | |
| Synthetic phosphopeptide corresponding to the sequence surrounding pSer792 of human raptor. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction