missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Raptor (Ser792) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585772
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: MCF-7, MCF-7(100 uM CCCP treatment for 2 hr). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Specifications
| Phospho-Raptor (Ser792) | |
| Polyclonal | |
| Unconjugated | |
| RPTOR | |
| 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Rap; RAPTOR; regulatory associated protein of MTOR complex 1; regulatory associated protein of MTOR, complex 1; regulatory-associated protein of mTOR; RGD1311784; RPTOR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57521 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.21 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8N122 | |
| RPTOR | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Ser792 of human Raptor. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction