missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RAD52 (Tyr104) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538702
This item is not returnable.
View return policy
Description
The protein encoded by this gene shares similarity with Saccharomyces cerevisiae Rad52, a protein important for DNA double-strand break repair and homologous recombination. This gene product was shown to bind single-stranded DNA ends, and mediate the DNA-DNA interaction necessary for the annealing of complementary DNA strands. It was also found to interact with DNA recombination protein RAD51, which suggested its role in RAD51 related DNA recombination and repair.
Specifications
| Phospho-RAD52 (Tyr104) | |
| Polyclonal | |
| Unconjugated | |
| RAD52 | |
| DNA repair protein RAD52 homolog, recombination protein RAD52, rhabdomyosarcoma antigen MU-RMS-40.23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19365, 5893 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P43351, P43352 | |
| RAD52 | |
| A synthetic phosphopeptide derived from human RAD52 around the phosphorylation site of Tyr104 (K-F-YP-V-G) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Non-human Primate | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction