missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RAD50 (Ser635) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5-106032
This item is not returnable.
View return policy
Description
Phospho-RAD50 (Ser635) Polyclonal antibody specifically detects Phospho-RAD50 (Ser635) in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Phospho-RAD50 (Ser635) | |
| Polyclonal | |
| Unconjugated | |
| RAD50 | |
| 153 kDa protein; DNA repair protein RAD50; hRAD50; hsRAD50; I79_004319; mRad50; Mrell; MRX complex DNA-binding subunit; N0872; NBSLD; RAD50; RAD50 2; RAD50 double strand break repair protein; RAD50 homolog (S. cerevisiae); RAD50 homolog, double strand break repair protein; RAD50 PEN; RAD502; RAD50-2; RAD50-3; Rad50l; YNL250W | |
| A synthesized peptide derived from human RAD50 around the phosphorylation site of Ser635. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70388, Q92878, Q9JIL8 | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 10111, 19360, 64012 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction