missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RAD17 (Ser656) Recombinant Rabbit Monoclonal Antibody (JE63-30)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556527
This item is not returnable.
View return policy
Description
Phospho-RAD17 (Ser656) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Rad17 is essential for sustained cell growth, maintenance of chromosomal stability, and ATR-dependent checkpoint activation upon DNA damage. It has a weak ATPase activity required for binding to chromatin. Rad17 also participates in the recruitment of the RAD1-RAD9-HUS1 complex and RHNO1 onto chromatin, and in CHEK1 activation. May also serve as a sensor of DNA replication progression, and may be involved in homologous recombination.
Specifications
| Phospho-RAD17 (Ser656) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75943, Q6NXW6 | |
| Rad17 | |
| Synthetic peptide corresponding to residues surrounding Ser656 of human Rad17 protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| JE63-30 | |
| Unconjugated | |
| Rad17 | |
| CCYC; cell cycle checkpoint protein (RAD17); cell cycle checkpoint protein RAD17; DNA damage checkpoint control protein RAD17; DNA repair exonuclease RAD17; FLJ41520; hRad17; MmRad24; R24L; RAD1 homolog; RAD17; RAD17 (Ser647); RAD17 checkpoint clamp loader component; RAD17 homolog; RAD17 homolog (S. pombe); Rad17 like protein; Rad17-like protein; Rad17p; RAD17SP; RAD24; RF-C activator 1 homolog; RF-C/activator 1 homolog; YOR368W | |
| Rabbit | |
| Protein A | |
| RUO | |
| 19356, 5884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction