Learn More
Invitrogen™ Phospho-PTP1B (Ser50) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5143754
Product Code. 17896768
  • £466.00 / 100µL
Description
The antibody detects a 50 kDa* doublet corresponding to PTP1B in western blots of calyculin A treated human A431 and Jurkat cells. This reactivity is observed as a weak singlet in control cells and is not observed after lambda phosphatase treatment.
PTP1B (PTPN1) is a tyrosine-protein phosphatase which acts as a regulator of endoplasmic reticulum unfolded protein response. It mediates dephosphorylation of EIF2AK3/PERK and in turn inactivates the protein kinase activity of EIF2AK3/PERK. PTP1B also plays a role in CKII- and p60c-src-induced signal transduction cascades. It regulates the EFNA5-EPHA3 signaling pathway which modulates cell reorganization and cell-cell repulsion. PTP1B also regulates the hepatocyte growth factor receptor signaling pathway through dephosphorylation of MET.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-PTP1B (Ser50)
Polyclonal
Unconjugated
PTPN1
CPTP1; EC 3.1.3.48; EGK_02300; hgnc9642; KIAA0283; protein tyrosine phosphatase 1B; protein tyrosine phosphatase non-receptor type 1; protein tyrosine phosphatase, non-receptor type 1; protein tyrosine phosphatase, placental; protein-protein-tyrosine phosphatase HA2; protein-tyrosine phosphatase 1B; Protein-tyrosine phosphatase HA2; PTN1; Ptp; PTP1B; PTP-1B; ptp1b protein; PTP-HA2; Ptpn1; RP5-1121H13.2; RPTPrho; tyrosine phosphatase 1b; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 1; wu:fk54h03
Rabbit
Antigen affinity chromatography
RUO
19246, 24697, 5770
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
P18031, P20417, P35821
PTPN1
PTP1B (Ser-50) synthetic peptide (coupled to carrier protein) corresponds to amino acids surrounding Ser-50 in human PTP1B. This sequence has significant homology to the conserved site in rat and mouse PTP1B, and has homology to conserved sites in TC-PTP and MPTP, but not other protein tyrosine phosphatase family members.
100 μL
Primary
Human, Mouse, Rat