missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PSEN2 (Ser330) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105831
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of Presenilin 2 only when phosphorylated at Ser330.
Alzheimer's disease (AD) patients with an inherited form of the disease carry mutations in the presenilin proteins (PSEN1 or PSEN2) or the amyloid precursor protein (APP). These disease-linked mutations result in increased production of the longer form of amyloid-beta (main component of amyloid deposits found in AD brains). Presenilins are postulated to regulate APP processing through their effects on gamma-secretase, an enzyme that cleaves APP. Also, it is thought that the presenilins are involved in the cleavage of the Notch receptor such that, they either directly regulate gamma-secretase activity, or themselves act are protease enzymes. Two alternatively spliced transcript variants encoding different isoforms of PSEN2 have been identified.
Specifications
| Phospho-PSEN2 (Ser330) | |
| Polyclonal | |
| Unconjugated | |
| PSEN2 | |
| AD3L; AD3LP; AD4; Ad4h; AD5; Alg3; ALG-3; Alzheimer disease 4; CMD1V; E5-1; presenilin 2; presenilin-2; Presenilin-2 CTF subunit; Presenilin-2 NTF subunit; PS2; Ps-2; PSEN 2; PSEN2; Psnl2; STM2; STM-2 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 19165, 5664, 81751 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88777, P49810, Q61144 | |
| PSEN2 | |
| A synthesized peptide derived from human PSEN2(Accession P49810), corresponding to amino acid residues around phosphorylated Ser330. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction