missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Protein APC (Ser2054) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599327
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of APC only when phosphorylated at Sersine 2054.
This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.
Specifications
| Phospho-Protein APC (Ser2054) | |
| Polyclonal | |
| Unconjugated | |
| Apc | |
| adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 11789, 324 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P25054, Q61315 | |
| Apc | |
| A synthesized peptide derived from human APC(Accession P25054), corresponding to amino acid residues around phosphorylated Ser2054. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction