missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Prohibitin (Tyr259) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537682
Code : NEW
Additional Details : Weight : 0.00910kg
Description
A suggested positive control for Western blot is MCF-7, PC12, 293 or NIH-3T3 cells.
Phospho-Prohibitin (Tyr259) Polyclonal antibody specifically detects Phospho-Prohibitin (Tyr259) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-Prohibitin (Tyr259) | |
Polyclonal | |
Unconjugated | |
PHB | |
BAP 32; Bap32; B-cell receptor-associated protein 32; epididymis luminal protein 215; epididymis secretory sperm binding protein Li 54e; fa18f07; fb09f05; HEL-215; HEL-S-54e; OTTHUMP00000165765; OTTHUMP00000165766; OTTHUMP00000220103; OTTHUMP00000220170; OTTHUMP00000220171; PHB; phb protein; PHB1; Prohibitin; sr:nyz128; wu:fa18f07; wu:fb09f05 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18673, 25344, 5245 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35232, P67778, P67779 | |
PHB | |
Peptide sequence around phosphorylation site of tyrosine 259 (I-T-Y(p)-L-P) derived from Human PHB. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |