missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PP2A alpha (Tyr307) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536874
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 38 kDa. Purity is >95% by SDS-PAGE.
Phospho-PP2A alpha (Tyr307) Polyclonal antibody specifically detects Phospho-PP2A alpha (Tyr307) in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Phospho-PP2A alpha (Tyr307) | |
Polyclonal | |
Unconjugated | |
PPP2CA | |
B55-GAMMA; hypothetical protein; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; PP2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; Pp2a1; PP2A-alpha; PP2Ac; PP2CA; PP2Calpha; Ppp2ca; PR52; PR55G; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A; protein phosphatase-2A-alpha; R75353; replication protein C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19052, 24672, 5515 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P63330, P63331, P67775 | |
PPP2CA | |
Synthetic phosphopeptide derived from human PP2A-alpha around the phosphorylation site of Tyrosine 307 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |